Klercide™ Neutral Detergent
Klercide™ Neutral Detergent is a low foaming non-ionic detergent solution that is a reliable residue cleaner formulated for cleanrooms.
- Sterile Neutral Formulation - pH-neutral, nonionic, non-corrosive, low-foaming detergent — with no extraneous chemicals, dyes or perfumes — is designed to be used directly on hard, non-porous surfaces.
- Shield with a check mark icon
- Powerful Cleaning Performance - Designed specifically for use in cleanrooms and other controlled areas, Klercide™ Neutral Detergent is highly effective against oils, sticky residues and spillages, and ideal for use in general cleaning protocols.
- Ready-to-Use Formats - Available in trigger spray and capped bottle formats — all designed to reduce handling and need for dilution, to streamline cleaning protocols and reduce cost-to-clean.
- Sterile Delivery System - Trigger-spray format features patented SteriShield™ Delivery System (SDS), providing sterility assurance of contents in bottle by forming a complete seal that prevents any air from (microbial or chemical contaminants) being drawn back into the bottle.
- Cleanroom-Grade Sterile Production - Solution is 0.2 micron-filtered and filled under unidirectional Grade A / ISO Class 7 airflow in a Grade B / ISO Class 5 cleanroom — and double-bagged to simplify movement into high-grade areas. Supported with Certificates of Analysis and Sterility with every batch.
For use in aseptic cleanrooms and associated areas, Klercide Neutral Detergent™ is used for cleaning small and large non-porous surfaces, general maintenance, and residue-removal. The product is low foaming, non-ionic with a neutral pH. Consistently and reliably remove soils and residues with a sterile neutral detergent formulated specifically for high-grade cleanroom environments and available in a variety of ready-to-use formats.
Neutral Detergent is 0.2 micron filtered, filled and capped in an ISO Class 7 cleanroom.
- Taille:4x1 gal
- Packaging:Bottle, double bagged, capped
- Sterility:Sterile
- Forme:Ready-to-use