Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® Wall Shelf Brackets, Fixed and Adjustable

VWR® Wall Shelf Brackets, Fixed and Adjustable

Adjustable Shelf Brackets, Snow white, 16" shelf, 16⅜", 6"
Catalog # CWM-0044-AD
Supplier:   VWR International
VWR® Wall Shelf Brackets, Fixed and Adjustable
Catalog # CWM-0044-AD
Supplier:  VWR International
VWR® Wall Shelf Brackets, Fixed and Adjustable
VWR® Wall Shelf Brackets, Fixed and Adjustable

Adjustable Shelf Brackets, Snow white, 16" shelf, 16⅜", 6"

Price
177,33 $
Log in for availability
Each
Product Details & Documents

Fixed and adjustable wall-mounted brackets.

  • Designed to suit a wide array of storage applications
  • Fixed brackets mount directly to the wall
  • Adjustable brackets are designed to mount to vertical wall supports (sold separately)

Fixed brackets features a flat top surface for simple shelf attachment. Adjustable brackets features a lipped top surface to provide increased security and stability when shelves are attached.

Ordering information: Fixed shelf brackets are supplied without mounting hardware. Adjustable brackets come with mounting hardware.

Specifications
  • Profondeur:
    16⅜"
  • Color:
    Snow white
  • Désignation:
    Adjustable Shelf Brackets
  • Hauteur:
    6"
  • For Use With:
    16" shelf