CrossLab Reciprocating Dissolution Tubes, Agilent Technologies
Outer tube, clear glass, calibrated, 50 ml, class B, for BIO-DIS
Catalog # CAAGG797247000
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Fournisseur #: G7972-47000
CrossLab Reciprocating Dissolution Tubes, Agilent Technologies
Catalog # CAAGG797247000
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Jump to:
Product Details & Documents
Several tube vessels are available for Apparatus 3 and 7.
- Reciprocating Cylinders for BIO-DIS and 400-DS
Ideal for automatic dissolution testing.
Spécifications
- Désignation:Outer tube, clear glass, calibrated, 50 ml, class B, for BIO-DIS