CrossLab Reciprocating Dissolution Tubes, Agilent Technologies | Avantor
CrossLab Reciprocating Dissolution Tubes, Agilent Technologies
Outer tube, clear glass, calibrated, 50 ml, class B, for BIO-DIS
Catalog # CAAGG797247000
Fournisseur #:  G7972-47000
CrossLab Reciprocating Dissolution Tubes, Agilent Technologies
Catalog # CAAGG797247000
CrossLab Reciprocating Dissolution Tubes, Agilent Technologies
CrossLab Reciprocating Dissolution Tubes, Agilent Technologies
Outer tube, clear glass, calibrated, 50 ml, class B, for BIO-DIS
Price
$770.00
Log in pour la disponibilité
Pack of 25
Product Details & Documents
Several tube vessels are available for Apparatus 3 and 7.
  • Reciprocating Cylinders for BIO-DIS and 400-DS
Ideal for automatic dissolution testing.
Spécifications
  • Désignation:
    Outer tube, clear glass, calibrated, 50 ml, class B, for BIO-DIS