QuickProbe GC/MS System Consumables, Agilent Technologies
QuickProbe GC/MS System Consumables, Agilent Technologies
QuickProbe probe, holder
  CAAGG397160200
Supplier #:  G3971-60200
QuickProbe GC/MS System Consumables, Agilent Technologies
  CAAGG397160200
QuickProbe GC/MS System Consumables, Agilent Technologies
QuickProbe GC/MS System Consumables, Agilent Technologies
QuickProbe probe, holder
$485.00
Each
QuickProbe GC/MS system consumables include insertion probes, columns, and specialized liners.
  • Unique fritted liner, 4 mm int.Ø
  • Two probe holders and three probe designs for unique sample types
  • Insertion probe round tip (recommended for liquids, dissolved samples, and tablets), 100/pk, used with QuickProbe probe holder
  • Insertion probe pocket tip (recommended for powders), 100/pk, used with QuickProbe probe holder
  • TSP solid probe vial (recommended for solid samples), 100/pk, used with TSP probe holder
  • QuickProbe DB-1HT, 0.25 mm × 0.1 μm × 2 m, for use in QuickProbe FastGC (high-temperature columns are highly recommended with the QuickProbe)
  • Designed for the QuickProbe GC/MS system
Agilent QuickProbe GC/MS system consumables include insertion probes, columns, and specialized liners. These supplies work with existing ferrules, gold seals, external split vent trap, and Agilent Gas Clean filters. Additional QuickProbe column phases can be ordered via the custom column process.
For Forensic use.
  • Désignation:
    QuickProbe probe, holder