QuickProbe GC/MS System Consumables, Agilent Technologies
QuickProbe TSP, probe holder
Catalog # CAAGG397120251
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Fournisseur #: G3971-20251
QuickProbe GC/MS System Consumables, Agilent Technologies
Catalog # CAAGG397120251
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Jump to:
Product Details & Documents
QuickProbe GC/MS system consumables include insertion probes, columns, and specialized liners.
- Unique fritted liner, 4 mm int.Ø
- Two probe holders and three probe designs for unique sample types
- Insertion probe round tip (recommended for liquids, dissolved samples, and tablets), 100/pk, used with QuickProbe probe holder
- Insertion probe pocket tip (recommended for powders), 100/pk, used with QuickProbe probe holder
- TSP solid probe vial (recommended for solid samples), 100/pk, used with TSP probe holder
- QuickProbe DB-1HT, 0.25 mm × 0.1 μm × 2 m, for use in QuickProbe FastGC (high-temperature columns are highly recommended with the QuickProbe)
- Designed for the QuickProbe GC/MS system
Agilent QuickProbe GC/MS system consumables include insertion probes, columns, and specialized liners. These supplies work with existing ferrules, gold seals, external split vent trap, and Agilent Gas Clean filters. Additional QuickProbe column phases can be ordered via the custom column process.
For Forensic use.
Spécifications
- Désignation:QuickProbe TSP, probe holder