SPEC Disk SPE, Agilent Technologies | Avantor
SPEC Disk SPE, Agilent Technologies
SPEC disks, C8, 47 mm, 47 mm
Catalog # CAAGA74702
Fournisseur #:  A74702
SPEC Disk SPE, Agilent Technologies
Catalog # CAAGA74702
SPEC Disk SPE, Agilent Technologies
SPEC Disk SPE, Agilent Technologies
SPEC disks, C8, 47 mm, 47 mm
Price
291,00 $
Log in pour la disponibilité
Pack of 24
Product Details & Documents
SPEC disk SPE delivers superior flow characteristics and trouble-free automation.
  • No loose sorbent means no channeling and better reproducibility
  • Uniform flow and extraction properties offer robust performance
  • Low elution volume affords excellent concentration of the analyte, improving sensitivity
SPEC disk SPE is a product providing faster sample processing, lower costs, and high-quality results. SPEC is manufactured with ultraclean polypropylene plastic and contains a monolithic sorbent bed with bonded silica infused on a glass fiber disk. For some applications, the elution volume is very small, eliminating the dry down and reconstitution steps.
Integrated filtration protects against clogging. SPEC is available in cartridge, 96-well plate, and disk formats in a wide range of chemistries for many applications.
Spécifications
  • Ø:
    47 mm
  • Désignation:
    SPEC disks, C8, 47 mm