Disintegration Baskets, Agilent Technologies
Basket, 1-tube, 44 mm OD, 10-mesh, 1905 µm
  CAAG37-3040
 :  37-3040
Disintegration Baskets, Agilent Technologies
  CAAG37-3040
 :  
Disintegration Baskets, Agilent Technologies
Disintegration Baskets, Agilent Technologies
Basket, 1-tube, 44 mm OD, 10-mesh, 1905 µm
Agilent offers disintegration baskets for the 100 automated disintegration apparatus.
  • Désignation:
    Basket, 1-tube, 44 mm OD, 10-mesh, 1905 µm