Disintegration Baskets, Agilent Technologies
Disintegration Baskets, Agilent Technologies
Basket, 3-tube, 38 mm OD, 10-mesh, 1905 µm, includes certificate
  CAAG37-3030V
Supplier #:  37-3030V
Disintegration Baskets, Agilent Technologies
  CAAG37-3030V
Disintegration Baskets, Agilent Technologies
Disintegration Baskets, Agilent Technologies
Basket, 3-tube, 38 mm OD, 10-mesh, 1905 µm, includes certificate
$1,530.00
Each
Agilent offers disintegration baskets for the 100 automated disintegration apparatus.
  • Désignation:
    Basket, 3-tube, 38 mm OD, 10-mesh, 1905 µm, includes certificate