Disintegration Baskets, Agilent Technologies | Avantor
Disintegration Baskets, Agilent Technologies
Basket, 6-tube, 10-mesh, 1905 µm (USP), includes certificate
Catalog # CAAG37-3001V
Fournisseur #:  37-3001V
Disintegration Baskets, Agilent Technologies
Catalog # CAAG37-3001V
Disintegration Baskets, Agilent Technologies
Disintegration Baskets, Agilent Technologies
Basket, 6-tube, 10-mesh, 1905 µm (USP), includes certificate
Price
$989.00
Log in pour la disponibilité
Each
Product Details & Documents
Agilent offers disintegration baskets for the 100 automated disintegration apparatus.
Spécifications
  • Désignation:
    Basket, 6-tube, 10-mesh, 1905 µm (USP), includes certificate