Disintegration Baskets, Agilent Technologies
Disintegration Baskets, Agilent Technologies
Basket, 6-tube, 20-mesh, 864 µm
Catalog # CAAG37-2010
Supplier #:  37-2010
Disintegration Baskets, Agilent Technologies
Catalog # CAAG37-2010
Disintegration Baskets, Agilent Technologies
Disintegration Baskets, Agilent Technologies
Basket, 6-tube, 20-mesh, 864 µm
Price
$873.00
Log in pour la disponibilité
Each
Product Details & Documents
Agilent offers disintegration baskets for the 100 automated disintegration apparatus.
Spécifications
  • Désignation:
    Basket, 6-tube, 20-mesh, 864 µm