CrossLab Reciprocating Dissolution Holders, Agilent Technologies | Avantor
CrossLab Reciprocating Dissolution Holders, Agilent Technologies
Basket,Mesh End, 20 µm, for 400-DS
Catalog # CAAG33-9046
Fournisseur #:  33-9046
CrossLab Reciprocating Dissolution Holders, Agilent Technologies
Catalog # CAAG33-9046
CrossLab Reciprocating Dissolution Holders, Agilent Technologies
CrossLab Reciprocating Dissolution Holders, Agilent Technologies
Basket,Mesh End, 20 µm, for 400-DS
Price
$1,930.00
Log in pour la disponibilité
Each
Product Details & Documents
Agilent can provide many different dosage form holders for the Apparatus 7.
  • Accessories for the reciprocating holder apparatus 7
  • Simulates the biorelevant conditions for the skin, gastrointestinal tract and tissue required for transdermal, oral drugs and implants with biorelevant temperature agitation rates and retention times
  • Accommodates a variety of dosage form holders including reciprocating disks, angled disks, spring holders, stent holders, and pointed rods
  • Supports typical volume configurations of 5 to 300 ml, depending on instrument
Ideal for automatic dissolution testing of dosage forms that require a change of media, a smaller volume, or more vigorous agitation. The Agilent 400-DS dissolution apparatus 7 enables of small-volume drug-release testing of medical devices such as drug eluting stents (DES).
Spécifications
  • Désignation:
    Basket,Mesh End, 20 µm, for 400-DS