Dissolution Conversion Kits, Agilent Technologies | Avantor
Dissolution Conversion Kits, Agilent Technologies
Conversion Kit, from 300 ml outer tubes to 1000 ml, for BIO-DIS
Catalog # CAAG27-6105
Fournisseur #:  27-6105
Dissolution Conversion Kits, Agilent Technologies
Catalog # CAAG27-6105
Dissolution Conversion Kits, Agilent Technologies
Dissolution Conversion Kits, Agilent Technologies
Conversion Kit, from 300 ml outer tubes to 1000 ml, for BIO-DIS
Price
$17,870.00
Log in pour la disponibilité
Kit of 1
Product Details & Documents
Agilent can also support small-volume dissolution with the 708-DS dissolution apparatus.
  • Special conversion kits: available in sets of 6 and 8, also available for the Chinese Pharmacopeia (CP) 250 ml vessel
  • Conversion kits are available to upgrade your 705-DS and 7000 and 7010 models to TruCenter vessels
The Agilent 708-DS dissolution apparatus can also support small-volume dissolution. Each position will require a conversion kit to hold a 100 or 200 ml vessel in position. The conversion kits are designed to be used with either mini paddles, mini baskets, or enhancer cells.
Easy-to-install conversion kits: the 708-DS is designed to accept 100 and 200 ml vessels, the 250 ml Chinese Pharmacopoeia vessel, and standard 1 L vessels. The 2 L model can incorporate 1 and 2 L vessels as well as smaller volume vessels with conversion kits.
Spécifications
  • Désignation:
    Conversion Kit, from 300 ml outer tubes to 1000 ml, for BIO-DIS