CrossLab Dissolution Paddles, Agilent Technologies
Paddle, electropolished stainless steel, 14.5", for VK7030
Catalog # CAAG14-3590
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Fournisseur #: 14-3590
CrossLab Dissolution Paddles, Agilent Technologies
Catalog # CAAG14-3590
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Jump to:
Product Details & Documents
Available in stainless steel, PTFE-coated, or PEEK in a range of sizes.
- All paddles are serialized and designed to meet USP specifications for Apparatus 2
- Select from electropolished stainless steel, PTFE-coated, or the biocompatible, chemically inert PEEK paddles in a range of sizes
- Additionally, mini and mega paddles for small- and large-volume dissolution testing are available
All paddles are serialized and designed to meet USP specifications for USP Apparatus 2 including an individual CoC as standard. Paddles are available in stainless steel, PTFE-coated, or PEEK in a range of standard, mega, or mini sizes.
Certifications: Each paddle includes an individual Certificate of Conformance as standard.
Spécifications
- Désignation:Paddle, electropolished stainless steel, 14.5", for VK7030
- Certification:Yes
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.