CrossLab Dissolution Shafts, Agilent Technologies
Basket shaft, USP 3‐clip, 14.5", for VK7025/7030
CAAG13-3615
: 13-3615
Shafts of different materials and lengths, individually serialized to meet standard USP design.
- Electropolished stainless steel, PTFE-coated, gold-plated or mini basket shafts available
- Individually serialized
- Three-clip shafts to meet standard USP design
- O-ring shafts for automated systems
- Conical shafts to support air bubble dispersion
- A range of basket shaft lengths are available to accommodate various models and volume configurations
Agilent provides stainless steel, PTFE-coated, gold-plated, or mini shafts that are individually serialized. Several basket and paddle shaft lengths and options are available to meet standard USP design.
- Désignation:Basket shaft, USP 3‐clip, 14.5", for VK7025/7030
- Certification:Yes