CrossLab Dissolution Paddles, Agilent Technologies
Paddle, PTFE-coated, 24", for 705-DS; 708-DS; 709-DS; VK7000
Catalog # CAAG13-3596
Fournisseur #: 13-3596
Jump to:
Discontinued
Product Details & Documents
Available in stainless steel, PTFE-coated, or PEEK in a range of sizes.
- All paddles are serialized and designed to meet USP specifications for Apparatus 2
- Select from electropolished stainless steel, PTFE-coated, or the biocompatible, chemically inert PEEK paddles in a range of sizes
- Additionally, mini and mega paddles for small- and large-volume dissolution testing are available
All paddles are serialized and designed to meet USP specifications for USP Apparatus 2 including an individual CoC as standard. Paddles are available in stainless steel, PTFE-coated, or PEEK in a range of standard, mega, or mini sizes.
Certifications: Each paddle includes an individual Certificate of Conformance as standard.
Spécifications
- Désignation:Paddle, PTFE-coated, 24", for 705-DS; 708-DS; 709-DS; VK7000
- Certification:Yes