Dissolution Evaporation Covers, Agilent Technologies
EaseAlign Evaporation cover, for use with 100/200 ml vessel, for 705-DS, VK7000,7010
Catalog # CAAG12-6315
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Fournisseur #: 12-6315
Dissolution Evaporation Covers, Agilent Technologies
Catalog # CAAG12-6315
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Jump to:
Product Details & Documents
A variety of evaporation covers are available to fit your apparatus and vessels.
- The evaporation covers automatically travel with the apparatus driveunit using either the alignment posts or DDM assemblies
- The standard evaporation cover on the 708-DS is designed to allow less than 1% evaporative loss over a 24-hour period under normal operating conditions
- Agilent also offers other low loss covers that can reduce evaporative loss even further
A variety of evaporation covers are available to fit your apparatus and vessels. Evaporation plugs are used to minimize evaporation by sealing unused ports in the evaporation covers.
Spécifications
- Désignation:EaseAlign Evaporation cover, for use with 100/200 ml vessel, for 705-DS, VK7000,7010
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.