Dissolution Evaporation Covers, Agilent Technologies
EaseAlign Evaporation cover, low actinic - red, with DDM opening, for 705-DS, VK7000,7010
Catalog # CAAG12-6311
 :  12-6311
Dissolution Evaporation Covers, Agilent Technologies
Catalog # CAAG12-6311
Dissolution Evaporation Covers, Agilent Technologies
Dissolution Evaporation Covers, Agilent Technologies
EaseAlign Evaporation cover, low actinic - red, with DDM opening, for 705-DS, VK7000,7010
Price
224,00 $
Log in pour la disponibilité
Each
Product Details & Documents
A variety of evaporation covers are available to fit your apparatus and vessels.
  • The evaporation covers automatically travel with the apparatus driveunit using either the alignment posts or DDM assemblies
  • The standard evaporation cover on the 708-DS is designed to allow less than 1% evaporative loss over a 24-hour period under normal operating conditions
  • Agilent also offers other low loss covers that can reduce evaporative loss even further
A variety of evaporation covers are available to fit your apparatus and vessels. Evaporation plugs are used to minimize evaporation by sealing unused ports in the evaporation covers.
Spécifications
  • Désignation:
    EaseAlign Evaporation cover, low actinic - red, with DDM opening, for 705-DS, VK7000,7010