Dissolution Centering Rings, Agilent Technologies | Avantor
Dissolution Centering Rings, Agilent Technologies
TruCenter Alignment Plate
Catalog # CAAG12-6115
Fournisseur #:  12-6115
Dissolution Centering Rings, Agilent Technologies
Catalog # CAAG12-6115
Supplier:  
Dissolution Centering Rings, Agilent Technologies
Dissolution Centering Rings, Agilent Technologies
TruCenter Alignment Plate
Discontinued
Product Details & Documents
Centering rings are available in various types and sizes.
  • Centering rings are available for the EaseAlign vessels of the legacy 705-DS, 7000, and 7010 dissolution apparatus
The correct positioning of vessels is critical for dissolution testing. Some dissolution vessels are held in place by a centering ring, which are available in various types and sizes.
Spécifications
  • Désignation:
    TruCenter Alignment Plate