Dissolution Centering Rings, Agilent Technologies
TruCenter Alignment Plate
Catalog # CAAG12-6115
Fournisseur #: 12-6115
Jump to:
Discontinued
Product Details & Documents
Centering rings are available in various types and sizes.
- Centering rings are available for the EaseAlign vessels of the legacy 705-DS, 7000, and 7010 dissolution apparatus
The correct positioning of vessels is critical for dissolution testing. Some dissolution vessels are held in place by a centering ring, which are available in various types and sizes.
Spécifications
- Désignation:TruCenter Alignment Plate