Dissolution Centering Rings, Agilent Technologies
EaseAlign, Locating Pins 12/Pack
Catalog # CAAG12-6065
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
: 12-6065
Dissolution Centering Rings, Agilent Technologies
Catalog # CAAG12-6065
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Jump to:
Product Details & Documents
Centering rings are available in various types and sizes.
- Centering rings are available for the EaseAlign vessels of the legacy 705-DS, 7000, and 7010 dissolution apparatus
The correct positioning of vessels is critical for dissolution testing. Some dissolution vessels are held in place by a centering ring, which are available in various types and sizes.
Specifications
- Désignation:EaseAlign, Locating Pins 12/Pack