Dissolution Centering Rings, Agilent Technologies | Avantor
Dissolution Centering Rings, Agilent Technologies
EaseAlign Vessel Retainer, for VK 7000/7010
Catalog # CAAG12-6050
Fournisseur #:  12-6050
Dissolution Centering Rings, Agilent Technologies
Catalog # CAAG12-6050
Dissolution Centering Rings, Agilent Technologies
Dissolution Centering Rings, Agilent Technologies
EaseAlign Vessel Retainer, for VK 7000/7010
Price
$337.00
Log in pour la disponibilité
Each
Product Details & Documents
Centering rings are available in various types and sizes.
  • Centering rings are available for the EaseAlign vessels of the legacy 705-DS, 7000, and 7010 dissolution apparatus
The correct positioning of vessels is critical for dissolution testing. Some dissolution vessels are held in place by a centering ring, which are available in various types and sizes.
Spécifications
  • Désignation:
    EaseAlign Vessel Retainer, for VK 7000/7010