Dissolution Centering Rings, Agilent Technologies
EaseAlign Vessel Retainer, for VK 7000/7010
Catalog # CAAG12-6050
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Fournisseur #: 12-6050
Dissolution Centering Rings, Agilent Technologies
Catalog # CAAG12-6050
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Jump to:
Product Details & Documents
Centering rings are available in various types and sizes.
- Centering rings are available for the EaseAlign vessels of the legacy 705-DS, 7000, and 7010 dissolution apparatus
The correct positioning of vessels is critical for dissolution testing. Some dissolution vessels are held in place by a centering ring, which are available in various types and sizes.
Spécifications
- Désignation:EaseAlign Vessel Retainer, for VK 7000/7010
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.