Dissolution Sinkers, Agilent Technologies | Avantor
Dissolution Sinkers, Agilent Technologies
Sinker basket, Japanese Pharmacopoeia (JP), stainless steel
Catalog # CAAG12-3070
Fournisseur #:  12-3070
Dissolution Sinkers, Agilent Technologies
Catalog # CAAG12-3070
Dissolution Sinkers, Agilent Technologies
Dissolution Sinkers, Agilent Technologies
Sinker basket, Japanese Pharmacopoeia (JP), stainless steel
Price
$518.00
Log in pour la disponibilité
Each
Product Details & Documents
A variety of dissolution sinkers are available depending on your dosage form.
  • Capsule wire, weights, and sinker baskets are used to retain a dosage form at the bottom of the vessel for testing when the dosage form is buoyant
  • A variety of options are available
  • The flexible capsule wire meets USP requirements and has a 0.032" diameter
  • Sinker baskets are offered in a variety of mesh sizes and made of stainless steel with press-on caps, using O-rings to seal the basket
  • Japanese Pharmacopoeia (JP) sinker baskets are available as an alternative
  • Three-prong capsule weights fit traditional capsule sizes one through three, and are sold individually
Capsule wire, weights, and sinker baskets are used to retain a solid dosage form at the bottom of the vessel for testing when the dosage form is buoyant. Depending on your dosage form, a variety of options are available.
Spécifications
  • Désignation:
    Sinker basket, Japanese Pharmacopoeia (JP), stainless steel
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.