Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Mopec Harmony Tissue Marking Dyes, Mortech | Avantor

Mopec Harmony Tissue Marking Dyes, Mortech

Orange, 250 ml (8 oz.)
Catalog # 76549-718
Supplier:   Mopec
Fournisseur #:  BG107-250
Mopec Harmony Tissue Marking Dyes, Mortech
Catalog # 76549-718
Supplier:  Mopec
CAS Number:  
Mopec Harmony Tissue Marking Dyes, Mortech
Mopec Harmony Tissue Marking Dyes, Mortech

Orange, 250 ml (8 oz.)

CAS Number:  
Price
$229.64
Log in pour la disponibilité
Each
Product Details & Documents

Mopec’s range of Harmony tissue marking dyes are used to permanently mark surgical tissue margins in Histological specimens. Marking dyes are available in nine colors, with a standardized viscosity over the range. This ensures that there is less variation in viscosity from one color to another as compared to competitor products and allows the easy marking of both small and large specimens with any chosen color.

  • Emulsified pigments are blended perfectly with an aqueous base to provide a consistent and high-quality product every time
  • Chemical and heat resistance to formalin, alcohols, xylene and wax provides clear visibility both prior and after tissue processing
  • Dries in 3 minutes, improving laboratory workflow at the grossing station
  • Nonflammable
  • Available in either 60 ml or 250 ml round neck, induction sealed bottles with a standard flip top lid

Harmony has also been purposely engineered to reduce in viscosity during application, giving users an easy to apply dye without any dripping or running and ensures an even coverage of dye on the specimen.

High concentrations of pigment in the dyes increase their durability and allow diagnostic specimens to be more easily marked. Harmony coats specimens with minimal penetration of the tissue surface, whilst its strong adhesion qualities mean the marking dye will not interfere with diagnostic interpretations, making it ideal for radical prostatectomies.

Spécifications

Spécifications

  • Color:
    Orange
  • Taille:
    250 ml (8 oz.)

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.