Paraffin Tapper | Avantor
Paraffin Tapper
Stainless steel paraffin tapper, 22×22×1.5 mm
Catalog # 76549-700
Supplier:   Mopec
Fournisseur #:  BG031
Paraffin Tapper
Catalog # 76549-700
Supplier:  Mopec
Paraffin Tapper
Paraffin Tapper
Stainless steel paraffin tapper, 22×22×1.5 mm
Price
$99.65
Log in pour la disponibilité
Each
Product Details & Documents
Designed to flatten tissue during the histology embedding process.
  • Stainless steel construction for durability and corrosion resistance
  • Ergonomic handle for comfortable, precise operation
  • Easy to clean and maintain for long-term use
  • Essential tool for pathology and histology workflows
Constructed from durable, laboratory-grade stainless steel, the Mopec Paraffin Tapper offers long-lasting performance and resistance to corrosion. Its ergonomic handle ensures a comfortable, controlled grip, reducing hand fatigue during repetitive use. The balanced design provides excellent control, making it ideal for use in histology, pathology, and research laboratories.
Spécifications
  • Désignation:
    Stainless steel paraffin tapper, 22×22×1.5 mm