Knife Sharpener, Variable Speed, Mortech
Knife Sharpener, Variable Speed, Mortech
110 V
Catalog # 76549-332
Supplier:   Mopec
Supplier #:  AH032
Knife Sharpener, Variable Speed, Mortech
Catalog # 76549-332
Supplier:  Mopec
Knife Sharpener, Variable Speed, Mortech
Knife Sharpener, Variable Speed, Mortech
110 V
Price
4 894,32 $
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This variable speed sharpener creates a precise angle and a consistent, exact bevel by sharpening, thinning, and honing which results in a stronger cutting edge. The honing wheels are mounted on parallel shafts that rotate in opposite directions.
  • Stainless steel case
Available in US and International voltage.
Ordering information: Replacement honing wheels available (See model AH034).
Specifications
  • Désignation:
    110 V