Liston Bone Cutting Forceps, Angled, Mortech
Effective bone cutting instrument for use during autopsies and post-mortem dissection. Sharp tips perform a clean extraction.
- Stainless steel
- Smooth grip
Available in a variety of lengths.
- Longueur:8.5"
- Curvature:Angled