Bone Chisel, Brunetti’s, Mortech
Right-hand handle, 11"
  76549-214
 :   Mopec
 :  AG015
Bone Chisel, Brunetti’s, Mortech
  76549-214
 :  Mopec
Bone Chisel, Brunetti’s, Mortech
Bone Chisel, Brunetti’s, Mortech
Right-hand handle, 11"
$299.22
Each
The Brunetti's chisels are specifically designed to score, cut, scrape, clean and sculpt around the spinal cord.
  • Standard grade stainless steel
Available in right handed and left handed options.
  • Désignation:
    Right-hand handle
  • Longueur:
    11"