Forceps, Russian Tissue, Mortech
Premium, 10"
  76548-834
 :   Mopec
 :  AB131
Forceps, Russian Tissue, Mortech
  76548-834
 :  Mopec
Forceps, Russian Tissue, Mortech
Forceps, Russian Tissue, Mortech
Premium, 10"
$266.80
Each
Rounded perimeter multi-tooth tip allows for maximum tissue grip. Variety of lengths and grades.
  • Stainless steel
  • Ribbed grip
  • Rounded multi-toothed tips
Available in standard or premium grade.
  • Longueur:
    10"
  • Grade:
    Premium