Forceps, Russian Tissue, Mortech | Avantor
Forceps, Russian Tissue, Mortech
Premium, 6"
Catalog # 76548-830
Supplier:   Mopec
Fournisseur #:  AB129
Forceps, Russian Tissue, Mortech
Catalog # 76548-830
Supplier:  Mopec
Forceps, Russian Tissue, Mortech
Forceps, Russian Tissue, Mortech
Premium, 6"
Price
$152.03
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Rounded perimeter multi-tooth tip allows for maximum tissue grip. Variety of lengths and grades.
  • Stainless steel
  • Ribbed grip
  • Rounded multi-toothed tips
Available in standard or premium grade.
Spécifications
  • Longueur:
    6"
  • Grade:
    Premium