Forceps, Ergonomic, Mortech
7", Smooth
Catalog # 76548-728
Supplier:   Mopec
 :  AB054
Forceps, Ergonomic, Mortech
Catalog # 76548-728
Supplier:  Mopec
Forceps, Ergonomic, Mortech
Forceps, Ergonomic, Mortech
7", Smooth
Price
191,97 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Unique perforated paddle handles for secure slip-free grip. Fingertip aligned perforations offer precise maneuvering. Extremely lightweight and versatile for a wide variety of tissue specimens. Can be cleaned with any laboratory cleaner or disinfectant.
  • Stainless steel
  • Broad grip
Available with smooth or serrated tips.
Spécifications
  • Longueur:
    7"
  • Tip Style:
    Smooth