Forceps, Russian Tissue, Mortech
Rounded perimeter multi-tooth tip allows for maximum tissue grip. Variety of lengths and grades.
- Stainless steel
- Ribbed grip
- Rounded multi-toothed tips
Available in standard or premium grade.
- Longueur:8"
- Grade:Standard