Scissors, Titanium, Iris, Mortech | Avantor
Scissors, Titanium, Iris, Mortech
4", Curved
Catalog # 76548-624
Supplier:   Mopec
Fournisseur #:  AA708
Scissors, Titanium, Iris, Mortech
Catalog # 76548-624
Supplier:  Mopec
Scissors, Titanium, Iris, Mortech
Scissors, Titanium, Iris, Mortech
4", Curved
Price
$351.60
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Titanium scissors weigh approximately 25% less than stainless steel, making them the first choice for many technicians. Titanium alloy instruments keep edges sharp longer than traditional instruments. More stain resistant than stainless steel. Helpful in cutting into the facial region.
  • Standard grade titanium stainless steel
Available with straight or curved edges. Do not use bleach or other caustic chemicals.
Spécifications
  • Longueur:
    4"
  • Curvature:
    Curved