Scissors, Disposable, Mortech | Avantor
Scissors, Disposable, Mortech
5.25", Sharp/Sharp
Catalog # 76548-606
Supplier:   Mopec
Fournisseur #:  AA603
Scissors, Disposable, Mortech
Catalog # 76548-606
Supplier:  Mopec
Scissors, Disposable, Mortech
Scissors, Disposable, Mortech
5.25", Sharp/Sharp
Price
$104.64
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These disposable scissors are highly recommended for post-mortem infectious cases.
  • Straight cutting edge
  • Not autoclavable
  • Single use only
Package of 12. Available in multiple lengths and tips.
Spécifications
  • Longueur:
    5.25"
  • Blade Type:
    Sharp/Sharp