Scissors, Disposable, Mortech
Scissors, Disposable, Mortech
4.5", Sharp/Blunt
Catalog # 76548-604
Supplier:   Mopec
Supplier #:  AA602
Scissors, Disposable, Mortech
Catalog # 76548-604
Supplier:  Mopec
Scissors, Disposable, Mortech
Scissors, Disposable, Mortech
4.5", Sharp/Blunt
Price
64,72 $
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These disposable scissors are highly recommended for post-mortem infectious cases.
  • Straight cutting edge
  • Not autoclavable
  • Single use only
Package of 12. Available in multiple lengths and tips.
Specifications
  • Longueur:
    4.5"
  • Blade Type:
    Sharp/Blunt