Scissors, Disposable, Mortech
4.5", Sharp/Sharp
  76548-602
 :   Mopec
 :  AA601
Scissors, Disposable, Mortech
  76548-602
 :  Mopec
Scissors, Disposable, Mortech
Scissors, Disposable, Mortech
4.5", Sharp/Sharp
$64.72
Each
These disposable scissors are highly recommended for post-mortem infectious cases.
  • Straight cutting edge
  • Not autoclavable
  • Single use only
Package of 12. Available in multiple lengths and tips.
  • Longueur:
    4.5"
  • Blade Type:
    Sharp/Sharp