Scissors, SuperCut Iris, Mortech | Avantor
Scissors, SuperCut Iris, Mortech
4.5", Straight
Catalog # 76548-588
Supplier:   Mopec
Fournisseur #:  AA528
Scissors, SuperCut Iris, Mortech
Catalog # 76548-588
Supplier:  Mopec
Scissors, SuperCut Iris, Mortech
Scissors, SuperCut Iris, Mortech
4.5", Straight
Price
$493.78
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
SuperCut Scissors are tempered and hardened for a longer, sharper life over ordinary steel. The SuperCut blade has a specially designed sharp end. Also feature rounded and polished finger rings.
  • Stainless steel
  • Razor sharp cutting edge
Available with straight or curved edges.
Spécifications
  • Longueur:
    4.5"
  • Curvature:
    Straight