Scissors, Iris Dissection, Mortech | Avantor
Scissors, Iris Dissection, Mortech
4", Curved
Catalog # 76548-486
Supplier:   Mopec
Fournisseur #:  AA147
Scissors, Iris Dissection, Mortech
Catalog # 76548-486
Supplier:  Mopec
Scissors, Iris Dissection, Mortech
Scissors, Iris Dissection, Mortech
4", Curved
Price
$418.92
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Also referred to as the 'Ribbon Scissor' because of the unique and finger-friendly, comfort handle. With extra wide finger rings offers comfortable post-mortem dissection. This precise and delicate scissor is made of the highest quality German stainless steel. Extra sharp tips allow for intricate work.
  • High grade stainless steel
  • Extra sharp tips
Available with straight or curved tips.
Spécifications
  • Longueur:
    4"
  • Curvature:
    Curved