Scissors, Iris, Angled, Sharp/Probe, Mortech | Avantor
Scissors, Iris, Angled, Sharp/Probe, Mortech
Premium Grade, 4.5"
Catalog # 76548-462
Supplier:   Mopec
Fournisseur #:  AA134
Scissors, Iris, Angled, Sharp/Probe, Mortech
Catalog # 76548-462
Supplier:  Mopec
Scissors, Iris, Angled, Sharp/Probe, Mortech
Scissors, Iris, Angled, Sharp/Probe, Mortech
Premium Grade, 4.5"
Price
$92.16
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal for small, precise, post-mortem dissections.
  • Angular end cutting edge
  • Sharp/probe point
Available in standard or premium grade.
Spécifications
  • Désignation:
    Premium Grade
  • Longueur:
    4.5"
  • Cat. No.:
    76548-462