Scissors, Mayo Dissection, Straight, Mortech | Avantor
Scissors, Mayo Dissection, Straight, Mortech
Premium Grade, 6.75"
Catalog # 76548-440
Supplier:   Mopec
Fournisseur #:  AA120
Scissors, Mayo Dissection, Straight, Mortech
Catalog # 76548-440
Supplier:  Mopec
Scissors, Mayo Dissection, Straight, Mortech
Scissors, Mayo Dissection, Straight, Mortech
Premium Grade, 6.75"
Price
$194.46
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Slightly blunt tips assure controlled, precise and safe post-mortem dissection with minimal damage to surrounding tissue.
  • Stainless steel
  • Straight cutting edge
  • Blunt tips
Available in standard or premium grade.
Spécifications
  • Désignation:
    Premium Grade
  • Longueur:
    6.75"