Scissors, Dissection, Sharp/Blunt, Curved, Mortech | Avantor
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Premium Grade, 6.5"
Catalog # 76548-424
Supplier:   Mopec
Fournisseur #:  AA112
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Catalog # 76548-424
Supplier:  Mopec
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Premium Grade, 6.5"
Price
$256.84
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These scissors are designed with one sharp tip, one blunt tip and a curved cutting edge. Ideal for a variety of post-mortem dissection needs.
  • Stainless steel
  • Curved cutting edge
  • Sharp/blunt tips
Available in standard or premium grade.
Spécifications
  • Désignation:
    Premium Grade
  • Longueur:
    6.5"