Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Premium Grade, 5.5"
  76548-422
 :   Mopec
Supplier #:  AA111
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
  76548-422
 :  Mopec
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Premium Grade, 5.5"
229,35 $
Each
These scissors are designed with one sharp tip, one blunt tip and a curved cutting edge. Ideal for a variety of post-mortem dissection needs.
  • Stainless steel
  • Curved cutting edge
  • Sharp/blunt tips
Available in standard or premium grade.
  • Désignation:
    Premium Grade
  • Longueur:
    5.5"