Scissors, Dissection, Sharp/Blunt, Straight, Mortech
Premium Grade, 5.5"
Catalog # 76548-416
Supplier:   Mopec
 :  AA108
Scissors, Dissection, Sharp/Blunt, Straight, Mortech
Catalog # 76548-416
Supplier:  Mopec
Scissors, Dissection, Sharp/Blunt, Straight, Mortech
Scissors, Dissection, Sharp/Blunt, Straight, Mortech
Premium Grade, 5.5"
Price
$199.45
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These scissors are designed with one sharp tip, one blunt tip and a straight cutting edge. Ideal for a variety of post-mortem dissection needs.
  • Stainless steel
  • Straight cutting edge
  • Sharp/blunt tips
Available in standard or premium grade.
Spécifications
  • Désignation:
    Premium Grade
  • Longueur:
    5.5"