Scissors, Iris, Angled, Sharp/Probe, Mortech
Standard Grade, 4.5"
  76548-374
 :   Mopec
 :  AA034
Scissors, Iris, Angled, Sharp/Probe, Mortech
  76548-374
 :  Mopec
Scissors, Iris, Angled, Sharp/Probe, Mortech
Scissors, Iris, Angled, Sharp/Probe, Mortech
Standard Grade, 4.5"
$67.22
Each
Ideal for small, precise, post-mortem dissections.
  • Angular end cutting edge
  • Sharp/probe point
Available in standard or premium grade.
  • Désignation:
    Standard Grade
  • Longueur:
    4.5"