Scissors, Enterotomy, Mortech
4.5", Right-handed
  76548-370
 :   Mopec
 :  AA032
Scissors, Enterotomy, Mortech
  76548-370
 :  Mopec
Scissors, Enterotomy, Mortech
Scissors, Enterotomy, Mortech
4.5", Right-handed
$52.25
Each
Excellent scissors for small cases.
  • Standard grade stainless steel
  • Angled end cutting edge
  • Lower probe point
Available in right handed and left handed options.
  • Longueur:
    4.5"
  • Handle:
    Right-handed