Scissors, Enterotomy, Mortech | Avantor
Scissors, Enterotomy, Mortech
3.5", Right-handed
Catalog # 76548-368
Supplier:   Mopec
Fournisseur #:  AA031
Scissors, Enterotomy, Mortech
Catalog # 76548-368
Supplier:  Mopec
Scissors, Enterotomy, Mortech
Scissors, Enterotomy, Mortech
3.5", Right-handed
Price
$42.29
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Excellent scissors for small cases.
  • Standard grade stainless steel
  • Angled end cutting edge
  • Lower probe point
Available in right handed and left handed options.
Spécifications
  • Longueur:
    3.5"
  • Handle:
    Right-handed