Scissors, Iris, Angled, Mortech
Scissors, Iris, Angled, Mortech
Standard Grade, 4.5"
Catalog # 76548-366
Supplier:   Mopec
Supplier #:  AA030
Scissors, Iris, Angled, Mortech
Catalog # 76548-366
Supplier:  Mopec
Scissors, Iris, Angled, Mortech
Scissors, Iris, Angled, Mortech
Standard Grade, 4.5"
Price
$69.65
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal for small, precise, post-mortem dissection.
  • Stainless steel
  • Angled end cutting edge
  • Sharp points
Available in standard or premium grade.
Specifications
  • Désignation:
    Standard Grade
  • Longueur:
    4.5"