Scissors, Iris, Curved, Mortech | Avantor
Scissors, Iris, Curved, Mortech
Standard Grade, 4.5"
Catalog # 76548-364
Supplier:   Mopec
Fournisseur #:  AA029
Scissors, Iris, Curved, Mortech
Catalog # 76548-364
Supplier:  Mopec
Scissors, Iris, Curved, Mortech
Scissors, Iris, Curved, Mortech
Standard Grade, 4.5"
Price
$72.21
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal for small, precise, post-mortem dissection.
  • Stainless steel
  • Curve cutting edge
  • Sharp points
Available in standard or premium grade.
Spécifications
  • Désignation:
    Standard Grade
  • Longueur:
    4.5"