Scissors, Metzenbaum, Straight, Mortech | Avantor
Scissors, Metzenbaum, Straight, Mortech
Standard Grade, 7"
Catalog # 76548-354
Supplier:   Mopec
Fournisseur #:  AA024
Scissors, Metzenbaum, Straight, Mortech
Catalog # 76548-354
Supplier:  Mopec
Scissors, Metzenbaum, Straight, Mortech
Scissors, Metzenbaum, Straight, Mortech
Standard Grade, 7"
Price
$67.22
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These scissors offer a unique combination of slim, blunt tips and thin, delicate shafts for post-mortem dissection with minimal damage to surrounding tissues. Variety of lengths and grades.
  • Stainless steel
  • Straight cutting edge
  • Narrow blunt points
Available in standard or premium grade.
Spécifications
  • Désignation:
    Standard Grade
  • Longueur:
    7"