Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Standard Grade, 4.5"
Catalog # 76548-326
Supplier:   Mopec
Supplier #:  AA010
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Catalog # 76548-326
Supplier:  Mopec
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Scissors, Dissection, Sharp/Blunt, Curved, Mortech
Standard Grade, 4.5"
Price
42,29 $
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These scissors are designed with one sharp tip, one blunt tip and a curved cutting edge. Ideal for a variety of post-mortem dissection needs.
  • Stainless steel
  • Curved cutting edge
  • Sharp/blunt tips
Available in standard or premium grade.
Specifications
  • Désignation:
    Standard Grade
  • Longueur:
    4.5"