Michels Solution , Statlab | Avantor
Michels Solution , Statlab
Michels Solution, 32 oz
Catalog # 76548-154
Supplier:   StatLab
Fournisseur #:  MS-32
Michels Solution , Statlab
Catalog # 76548-154
Supplier:  StatLab
CAS Number:  
Michels Solution , Statlab
Michels Solution , Statlab
Michels Solution, 32 oz
CAS Number:  
Price
$115.01
Log in pour la disponibilité
Each (32oz)
Product Details & Documents
Michels Solution is used to preserve and transport specimens for examination in immunofluorescence studies.
  • While not a fixative, specimens can be held at room temperature for five days in transport medium before processing
  • This simple salt solution maintains pH, but does not kill most pathogens
  • Specimens received in transport medium should be washed in three changes of Michels Wash Solution (10 minutes for each wash)
  • Not suitable for FISH studies
Part of the StatLab line of formalin alternatives.
Manufactured in the USA
Spécifications
Spécifications
  • Désignation:
    Michels Solution
  • Taille:
    32 oz
Spécifications