Vee Gee Snap Cap Replacements
#1, 5 and 10 ml flasks (76546-110)
Catalog # 76546-716
Supplier: Heathrow Scientific
Supplier #: SC#1
Jump to:
Product Details & Documents
Replacement snap-caps.
- For SIBATA volumetric flasks
Sold by the each.
Specifications
- Taille du bouchon:#1
- Fits:5 and 10 ml flasks (76546-110)